Assay Details
Target Gene Details
Entrez Gene ID: | 3790 |
Gene Name: | potassium voltage-gated channel modifier subfamily S member 3 |
Gene Aliases: |
KV9.3 |
Location: |
Chr.2:17877847-17932961 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNS3 | NM_001282428.1 | NP_001269357.1 | ||
| NM_002252.4 | NP_002243.3 | |||
| XM_011532825.1 | XP_011531127.1 | |||
| AK225833.1 | ||||
| AK314451.1 | ||||
| BC004148.2 | AAH04148.1 | |||
| BC004987.1 | AAH04987.1 | |||
| BC015947.2 | AAH15947.1 | |||
| DA921825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891603 | Chr.2:17752895 - 18037489 on Build GRCh38 | Gain |
|
| nsv997502 | Chr.2:17160707 - 18147073 on Build GRCh38 | Gain |
|
| nsv1126541 | Chr.2:17878433 - 17878833 on Build GRCh38 | Deletion |
|
| esv3589949 | Chr.2:17755879 - 18035557 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map