Assay Details
Target Gene Details
Entrez Gene ID: | 22878 |
Gene Name: | trafficking protein particle complex 8 |
Gene Aliases: |
GSG1, HsT2706, KIAA1012, TRS85 |
Location: |
Chr.18:31829173-31943218 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAPPC8 | NM_014939.3 | NP_055754.2 | ||
| XM_005258233.2 | XP_005258290.1 | |||
| XM_005258234.2 | XP_005258291.1 | |||
| XM_005258235.3 | XP_005258292.1 | |||
| XM_011525884.2 | XP_011524186.1 | |||
| XM_017025615.1 | XP_016881104.1 | |||
| XM_017025616.1 | XP_016881105.1 | |||
| XM_017025617.1 | XP_016881106.1 | |||
| AB023229.1 | ||||
| AK001662.1 | ||||
| AL136749.1 | CAB66683.2 | |||
| BC127109.1 | ||||
| BC127110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893086 | Chr.18:31623572 - 31852661 on Build GRCh38 | Gain |
|
| nsv2247 | Chr.18:31790879 - 31835749 on Build GRCh38 | Deletion |
|
| esv3642136 | Chr.18:31801423 - 31902584 on Build GRCh38 | Gain |
|
| esv3642135 | Chr.18:31792111 - 31849505 on Build GRCh38 | Gain |
|
| nsv1057229 | Chr.18:31755212 - 31934302 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map