Assay Details
Target Gene Details
Entrez Gene ID: | 79966 |
Gene Name: | stearoyl-CoA desaturase 5 |
Gene Aliases: |
ACOD4, FADS4, HSCD5, SCD2, SCD4 |
Location: |
Chr.4:82629537-82798857 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCD5 | NM_001037582.2 | 5 | 2176 | NP_001032671.2 |
| AF389338.1 | 5 | 2091 | AAP31443.1 | |
| AL831891.1 | 3 | 1347 | CAD38567.1 | |
| BC048971.1 | 2 | 1270 | AAH48971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594744 | Chr.4:82570382 - 82837674 on Build GRCh38 | Gain |
|
| nsv829989 | Chr.4:82435486 - 82636734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map