Assay Details
Target Gene Details
Entrez Gene ID: | 51411 |
Gene Name: | bridging integrator 2 |
Gene Aliases: |
BRAP-1 |
Location: |
Chr.12:51281038-51324680 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BIN2 | NM_001290007.1 | NP_001276936.1 | ||
| NM_001290008.1 | NP_001276937.1 | |||
| NM_001290009.1 | NP_001276938.1 | |||
| NM_016293.3 | NP_057377.3 | |||
| XM_005268957.3 | XP_005269014.2 | |||
| XM_005268958.3 | XP_005269015.1 | |||
| XM_017019422.1 | XP_016874911.1 | |||
| AB032698.1 | BAA88108.1 | |||
| AF146531.1 | AAD54227.1 | |||
| AK000783.1 | BAA91376.1 | |||
| AK093638.1 | ||||
| AK225911.1 | ||||
| AK300211.1 | ||||
| AK314304.1 | ||||
| BC047686.1 | AAH47686.1 | |||
| CB996605.1 | ||||
| DB035315.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv558877 | Chr.12:51296967 - 51340167 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs151019240] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map