Assay Details
Target Gene Details
Entrez Gene ID: | 9882 |
Gene Name: | TBC1 domain family member 4 |
Gene Aliases: |
AS160, NIDDM5 |
Location: |
Chr.13:75284664-75482646 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D4 | NM_001286658.2 | 20 | 4178 | NP_001273587.1 |
| NM_001286659.2 | 19 | 4013 | NP_001273588.1 | |
| NM_014832.4 | 21 | 4202 | NP_055647.2 | |
| XM_005266603.1 | 20 | 4605 | XP_005266660.1 | |
| XM_005266605.2 | 21 | 3530 | XP_005266662.1 | |
| XM_006719903.3 | 21 | 3890 | XP_006719966.1 | |
| XM_011535331.1 | 20 | 4569 | XP_011533633.1 | |
| XM_017020882.1 | 19 | 3222 | XP_016876371.1 | |
| XM_017020883.1 | 18 | 3216 | XP_016876372.1 | |
| XM_017020884.1 | 9 | 2788 | XP_016876373.1 | |
| AB011175.1 | 22 | 4151 | ||
| AB449885.1 | 22 | 3804 | ||
| AK300468.1 | 10 | 1711 | ||
| AK304091.1 | 9 | 1532 | ||
| BC151239.1 | 22 | 4143 | ||
| CR749622.1 | 11 | 1982 | CAH18416.1 | |
| FM207106.1 | 19 | 3612 | ||
| FM207107.1 | 20 | 3777 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain |
|
| esv3632746 | Chr.13:75281592 - 75326999 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map