Assay Details
Target Gene Details
Entrez Gene ID: | 2494 |
Gene Name: | nuclear receptor subfamily 5 group A member 2 |
Gene Aliases: |
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2 |
Location: |
Chr.1:200027602-200177424 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR5A2 | NM_001276464.1 | 4 | 738 | NP_001263393.1 |
| NM_003822.4 | 4 | 907 | NP_003813.1 | |
| NM_205860.2 | 5 | 1045 | NP_995582.1 | |
| XM_005245062.3 | 4 | 1172 | XP_005245119.1 | |
| XM_011509380.1 | 6 | 1633 | XP_011507682.1 | |
| XM_011509381.2 | 5 | 2497 | XP_011507683.1 | |
| XM_011509382.1 | 5 | 1495 | XP_011507684.1 | |
| XM_011509384.2 | 5 | 878 | XP_011507686.1 | |
| XM_017000904.1 | 4 | 750 | XP_016856393.1 | |
| AB019246.1 | 5 | 925 | BAA34092.1 | |
| AB307719.1 | 4 | 679 | ||
| AF049102.1 | 3 | 561 | AAD03248.1 | |
| AF124247.1 | 5 | 799 | AAD26565.1 | |
| AF146343.1 | 4 | 868 | AAD37378.1 | |
| AK304344.1 | 4 | 776 | ||
| AK304365.1 | 4 | 738 | ||
| AK308446.1 | 4 | 861 | ||
| AK316513.1 | 4 | 974 | ||
| BC118571.1 | 4 | 754 | ||
| BC118652.1 | 4 | 754 | ||
| HQ692857.1 | 4 | 661 | ||
| U80251.1 | 4 | 716 | AAC78727.1 | |
| U93553.1 | 5 | 1041 | AAD03155.1 |
More Information
Set Membership: |
|

Genomic Map