Assay Details
Target Gene Details
Entrez Gene ID: | 84842 |
Gene Name: | 4-hydroxyphenylpyruvate dioxygenase like |
Gene Aliases: |
4-HPPD-L, GLOXD1 |
Location: |
Chr.1:45326873-45328675 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPDL | NM_032756.2 | 1 | 1353 | NP_116145.1 |
| AI808699.1 | 1 | 485 | ||
| AK313714.1 | ||||
| BC007293.1 | 1 | 1321 | AAH07293.1 |
More Information
Set Membership: |
|

Genomic Map