Assay Details
Target Gene Details
Entrez Gene ID: | 2744 |
Gene Name: | glutaminase |
Gene Aliases: |
AAD20, GAC, GAM, GLS1, KGA |
Location: |
Chr.2:190880821-190965552 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLS | NM_001256310.1 | NP_001243239.1 | ||
| NM_014905.4 | NP_055720.3 | |||
| XM_005246467.3 | XP_005246524.1 | |||
| XM_006712435.1 | XP_006712498.1 | |||
| XM_017003829.1 | XP_016859318.1 | |||
| AB020645.1 | ||||
| AF097492.1 | AAF00088.1 | |||
| AF097495.1 | AAF00090.1 | |||
| AF158555.1 | AAD47056.1 | |||
| AF223943.1 | AAF33825.1 | |||
| AF327434.1 | AAG47842.1 | |||
| AK289747.1 | ||||
| BC038507.1 | AAH38507.2 | |||
| Y08264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3083 | Chr.2:190880106 - 190914039 on Build GRCh38 | Insertion |
|
| nsv584065 | Chr.2:190779811 - 191011175 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78219026] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map