Assay Details
Target Gene Details
Entrez Gene ID: | 8437 |
Gene Name: | RAS protein activator like 1 |
Gene Aliases: |
RASAL |
Location: |
Chr.12:113096515-113136239 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASAL1 | NM_001193520.1 | 10 | 1097 | NP_001180449.1 |
| NM_001193521.1 | 9 | 1088 | NP_001180450.1 | |
| NM_001301202.1 | 9 | 933 | NP_001288131.1 | |
| NM_004658.2 | 10 | 1112 | NP_004649.2 | |
| XM_005253950.4 | 10 | 1112 | XP_005254007.1 | |
| XM_006719641.3 | 10 | 951 | XP_006719704.1 | |
| XM_006719642.3 | 9 | 1084 | XP_006719705.1 | |
| XM_011538852.2 | 9 | 1084 | XP_011537154.1 | |
| XM_011538853.2 | 9 | 1084 | XP_011537155.1 | |
| XM_017020028.1 | 10 | 1112 | XP_016875517.1 | |
| XM_017020029.1 | 9 | 1084 | XP_016875518.1 | |
| XM_017020030.1 | 10 | 1112 | XP_016875519.1 | |
| XM_017020031.1 | 10 | 1112 | XP_016875520.1 | |
| AB208935.1 | 10 | 1082 | BAD92172.1 | |
| AF086713.1 | 10 | 1089 | AAD09006.1 | |
| AK294350.1 | 10 | 1112 | ||
| AK309562.1 | 9 | 1085 | ||
| AL136672.1 | 9 | 1029 | CAB66607.2 | |
| BC014420.1 | 9 | 979 | AAH14420.1 | |
| BC093724.1 | 9 | 838 | AAH93724.1 | |
| BC143261.1 | 9 | 838 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560266 | Chr.12:113084969 - 113136469 on Build GRCh38 | Loss |
|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| nsv560264 | Chr.12:113050137 - 113136469 on Build GRCh38 | Loss |
|
| dgv2884n54 | Chr.12:113115603 - 113166364 on Build GRCh38 | Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map