Assay Details
Target Gene Details
Entrez Gene ID: | 26000 |
Gene Name: | TBC1 domain family member 10B |
Gene Aliases: |
EPI64B, FP2461 |
Location: |
Chr.16:30357101-30370258 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D10B | NM_015527.3 | NP_056342.3 | ||
| XM_011545789.1 | XP_011544091.1 | |||
| XM_011545790.2 | XP_011544092.1 | |||
| AB449895.1 | ||||
| AF370370.1 | AAQ15206.1 | |||
| AK023192.1 | BAB14454.1 | |||
| BC063112.1 | AAH63112.1 | |||
| BC072453.1 | ||||
| BC093814.1 | AAH93814.1 | |||
| BC093816.1 | AAH93816.1 | |||
| BC098419.1 | AAH98419.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827612 | Chr.16:30360217 - 30418541 on Build GRCh38 | Gain |
|
| esv2758642 | Chr.16:30062634 - 30371959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map