Assay Details
Target Gene Details
Entrez Gene ID: | 222584 |
Gene Name: | family with sequence similarity 83 member B |
Gene Aliases: |
C6orf143 |
Location: |
Chr.6:54846323-54945099 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM83B | NM_001010872.2 | 2 | 498 | NP_001010872.1 |
| XM_006715022.3 | 2 | 527 | XP_006715085.1 | |
| XM_011514394.2 | 2 | 540 | XP_011512696.1 | |
| XM_011514395.2 | 2 | 1529 | XP_011512697.1 | |
| XM_017010478.1 | 2 | 524 | XP_016865967.1 | |
| AK055204.1 | 2 | 499 | BAB70873.1 | |
| BC101628.1 | 1 | 457 | AAI01629.1 | |
| BC112275.1 | 1 | 457 | AAI12276.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3609057 | Chr.6:54860092 - 54902029 on Build GRCh38 | Gain |
|
| esv2763553 | Chr.6:54823181 - 54955689 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map