Assay Details
Target Gene Details
Entrez Gene ID: | 1195 |
Gene Name: | CDC like kinase 1 |
Gene Aliases: |
CLK, CLK/STY, STY |
Location: |
Chr.2:200853009-200864744 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLK1 | NM_004071.3 | NP_004062.2 | ||
| NR_027855.1 | ||||
| NR_027856.1 | ||||
| AK289365.1 | ||||
| AK307301.1 | ||||
| BE241788.1 | ||||
| DA225472.1 |
More Information
Set Membership: |
|

Genomic Map