Assay Details
Target Gene Details
Entrez Gene ID: | 283 |
Gene Name: | angiogenin |
Gene Aliases: |
ALS9, HEL168, RAA1, RNASE4, RNASE5 |
Location: |
Chr.14:20684177-20694186 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANG | NM_001097577.2 | NP_001091046.1 | ||
| NM_001145.4 | NP_001136.1 | |||
| AK313989.1 | ||||
| AV646929.1 | ||||
| BC020704.1 | ||||
| BC054880.1 | AAH54880.1 | |||
| BC062698.1 | AAH62698.1 | |||
| BQ773604.1 | ||||
| CB963791.1 | ||||
| FJ236304.1 |
Target Gene Details
Entrez Gene ID: | 6038 |
Gene Name: | ribonuclease A family member 4 |
Gene Aliases: |
RAB1, RNS4 |
Location: |
Chr.14:20684213-20700602 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNASE4 | NM_001282192.1 | NP_001269121.1 | ||
| NM_001282193.1 | NP_001269122.1 | |||
| NM_002937.4 | NP_002928.1 | |||
| NM_194431.2 | NP_919412.1 | |||
| AK092422.1 | ||||
| BC015520.1 | AAH15520.1 | |||
| BG616902.1 | ||||
| BU594423.1 | ||||
| BX247985.1 | CAD62319.1 | |||
| BX248010.1 | CAD62338.1 | |||
| DA925725.1 | ||||
| DR003809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3633688 | Chr.14:20684511 - 20707081 on Build GRCh38 | Loss |
|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
| esv3633687 | Chr.14:20683687 - 20720960 on Build GRCh38 | Loss |
|
| nsv1203 | Chr.14:20652709 - 20697918 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map