Assay Details
Target Gene Details
Entrez Gene ID: | 128876 |
Gene Name: | family with sequence similarity 83 member C |
Gene Aliases: |
C20orf128, dJ614O4.7 |
Location: |
Chr.20:35285731-35292422 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM83C | NM_178468.5 | NP_848563.1 | ||
| AK091116.1 | ||||
| AK091138.1 | BAC03594.1 | |||
| BC113483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059168 | Chr.20:35217473 - 35319358 on Build GRCh38 | Loss |
|
| nsv954029 | Chr.20:35284398 - 35298997 on Build GRCh38 | Deletion |
|
| nsv585943 | Chr.20:35247970 - 35297352 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map