Assay Details
Target Gene Details
Entrez Gene ID: | 283987 |
Gene Name: | HID1 domain containing |
Gene Aliases: |
C17orf28, DMC1, HID-1 |
Location: |
Chr.17:74950742-74972805 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HID1 | NM_030630.2 | NP_085133.1 | ||
| XM_005257226.1 | XP_005257283.1 | |||
| AK074401.1 | BAB85070.1 | |||
| AK095565.1 | ||||
| AK304081.1 | ||||
| AL137556.1 | CAB70810.2 | |||
| BC032219.1 | AAH32219.2 | |||
| BC035372.1 | AAH35372.1 | |||
| DA336027.1 |
Target Gene Details
Entrez Gene ID: | 102723641 |
Gene Name: | HID1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.17:74970689-74975728 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HID1-AS1 | NR_110878.1 | |||
| CD515974.1 | ||||
| HY081987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv470601 | Chr.17:74931028 - 74982818 on Build GRCh38 | Loss |
|
| nsv576026 | Chr.17:74889500 - 74982818 on Build GRCh38 | Gain |
|
| nsv1056788 | Chr.17:74922211 - 75028689 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map