Assay Details
Target Gene Details
Entrez Gene ID: | 3431 |
Gene Name: | SP110 nuclear body protein |
Gene Aliases: |
IFI41, IFI75, IPR1, VODI |
Location: |
Chr.2:230165466-230225729 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP110 | NM_001185015.1 | 15 | 1805 | NP_001171944.1 |
| NM_004509.3 | 14 | 1769 | NP_004500.3 | |
| NM_004510.3 | 14 | 1769 | NP_004501.3 | |
| NM_080424.2 | 14 | 1769 | NP_536349.2 | |
| XM_005246525.3 | 15 | 1805 | XP_005246582.1 | |
| XM_006712487.3 | 15 | 1805 | XP_006712550.1 | |
| XM_006712489.3 | 15 | 1805 | XP_006712552.1 | |
| XM_011511088.2 | 15 | 1805 | XP_011509390.1 | |
| XM_011511089.2 | 14 | 1660 | XP_011509391.1 | |
| XM_011511090.2 | 14 | 1655 | XP_011509392.1 | |
| XM_011511091.2 | 15 | 1805 | XP_011509393.1 | |
| XM_011511092.2 | 13 | 1498 | XP_011509394.1 | |
| XM_017003968.1 | 16 | 1880 | XP_016859457.1 | |
| XM_017003969.1 | 14 | 1655 | XP_016859458.1 | |
| XM_017003970.1 | 13 | 1510 | XP_016859459.1 | |
| AF280094.1 | 14 | 1606 | AAF99318.1 | |
| AF280095.1 | 14 | 1606 | AAG09826.1 | |
| AK026488.1 | 14 | 1608 | ||
| AK301097.1 | 15 | 1805 | ||
| AL579921.3 | 2 | 278 | ||
| AL832300.1 | 13 | 1543 | ||
| BC019059.2 | 14 | 1624 | AAH19059.1 | |
| BG231996.1 | 2 | 314 | ||
| CA432211.1 | 2 | 331 | ||
| L22343.1 | 11 | 1090 | AAD13402.1 |
More Information
Set Membership: |
|

Genomic Map