Assay Details
Target Gene Details
Entrez Gene ID: | 283248 |
Gene Name: | REST corepressor 2 |
Gene Aliases: |
- |
Location: |
Chr.11:63911221-63916844 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCOR2 | NM_173587.3 | NP_775858.2 | ||
| XM_005273932.3 | XP_005273989.1 | |||
| AB593132.1 | ||||
| BC010608.1 | AAH10608.1 | |||
| BC023587.2 | AAH23587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555180 | Chr.11:63748280 - 63945767 on Build GRCh38 | Loss |
|
| nsv555181 | Chr.11:63872409 - 64015880 on Build GRCh38 | Loss |
|
| esv2660413 | Chr.11:63908673 - 63918220 on Build GRCh38 | Deletion |
|
| nsv951009 | Chr.11:63901229 - 63928128 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map