Assay Details
Target Gene Details
Entrez Gene ID: | 9855 |
Gene Name: | FERM, ARH/RhoGEF and pleckstrin domain protein 2 |
Gene Aliases: |
FIR, FRG, PLEKHC3 |
Location: |
Chr.2:241356249-241494842 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARP2 | NM_014808.3 | 27 | 3961 | NP_055623.1 |
| XM_011512233.1 | 28 | 3946 | XP_011510535.1 | |
| XM_011512234.1 | 28 | 3871 | XP_011510536.1 | |
| XM_011512235.1 | 29 | 4204 | XP_011510537.1 | |
| XM_011512236.1 | 28 | 4000 | XP_011510538.1 | |
| XM_017005416.1 | 29 | 4161 | XP_016860905.1 | |
| XM_017005417.1 | 28 | 4140 | XP_016860906.1 | |
| XM_017005418.1 | 25 | 3778 | XP_016860907.1 | |
| AB018336.1 | 27 | 3914 | ||
| AK127510.1 | 6 | 3192 | ||
| AL122052.1 | 6 | 1326 | CAB59185.1 | |
| AW014580.1 | 1 | 101 | ||
| HY312568.1 | 1 | 115 |
Target Gene Details
Entrez Gene ID: | 10494 |
Gene Name: | serine/threonine kinase 25 |
Gene Aliases: |
SOK1, YSK1 |
Location: |
Chr.2:241494707-241509730 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK25 | NM_001271977.1 | NP_001258906.1 | ||
| NM_001271978.1 | NP_001258907.1 | |||
| NM_001271979.1 | NP_001258908.1 | |||
| NM_001271980.1 | NP_001258909.1 | |||
| NM_001282305.1 | NP_001269234.1 | |||
| NM_001282306.1 | NP_001269235.1 | |||
| NM_001282307.1 | NP_001269236.1 | |||
| NM_001282308.1 | NP_001269237.1 | |||
| NM_006374.4 | NP_006365.2 | |||
| HY315908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759120 | Chr.2:241480540 - 241562243 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map