Assay Details
Target Gene Details
Entrez Gene ID: | 5304 |
Gene Name: | prolactin induced protein |
Gene Aliases: |
GCDFP-15, GCDFP15, GPIP4 |
Location: |
Chr.7:143132081-143139741 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIP | NM_002652.2 | NP_002643.1 | ||
| AK312227.1 | ||||
| BC010950.1 | AAH10950.1 | |||
| BC010951.1 | AAH10951.1 | |||
| CD723133.1 | ||||
| J03460.1 | AAA60091.1 | |||
| Y10179.1 | CAA71252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv11661n54 | Chr.7:143125117 - 143200731 on Build GRCh38 | Loss |
|
| esv3615300 | Chr.7:143097437 - 143200309 on Build GRCh38 | Loss |
|
| esv2675537 | Chr.7:143127749 - 143196814 on Build GRCh38 | Deletion |
|
| esv3542554 | Chr.7:143127408 - 143198705 on Build GRCh38 | Deletion |
|
| nsv608657 | Chr.7:143070653 - 143184447 on Build GRCh38 | Loss |
|
| dgv11662n54 | Chr.7:143130861 - 143174945 on Build GRCh38 | Loss |
|
| dgv6698n100 | Chr.7:143119834 - 143203251 on Build GRCh38 | Loss |
|
| esv2764005 | Chr.7:143117686 - 143205205 on Build GRCh38 | Loss |
|
| dgv432n21 | Chr.7:143095565 - 143208912 on Build GRCh38 | Loss |
|
| esv3615302 | Chr.7:143127752 - 143196816 on Build GRCh38 | Loss |
|
| dgv1977e212 | Chr.7:143123700 - 143202300 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map