Assay Details
Target Gene Details
Entrez Gene ID: | 119392 |
Gene Name: | SWI5 dependent homologous recombination repair protein 1 |
Gene Aliases: |
C10orf78, MEI5, MEIR5, bA373N18.1 |
Location: |
Chr.10:104122058-104126385 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SFR1 | NM_001002759.1 | NP_001002759.1 | ||
| NM_145247.4 | NP_660290.3 | |||
| XM_005269521.3 | 1 | 691 | XP_005269578.1 | |
| XM_017015672.1 | XP_016871161.1 | |||
| AK123954.1 | ||||
| AK291184.1 | 1 | 741 | ||
| BC020892.2 | AAH20892.2 | |||
| BC140835.1 | ||||
| BG942691.1 | ||||
| CD694866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831971 | Chr.10:104120100 - 104301309 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map