Assay Details
Target Gene Details
Entrez Gene ID: | 50835 |
Gene Name: | taste 2 receptor member 9 |
Gene Aliases: |
T2R9, TRB6 |
Location: |
Chr.12:10809094-10810168 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS2R9 | NM_023917.2 | 1 | 461 | NP_076406.1 |
| BC069066.1 | 1 | 368 | AAH69066.1 | |
| BC095519.1 | 1 | 462 | AAH95519.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3628543 | Chr.12:10720386 - 10881212 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map