Assay Details
Target Gene Details
Entrez Gene ID: | 6428 |
Gene Name: | serine and arginine rich splicing factor 3 |
Gene Aliases: |
SFRS3, SRp20 |
Location: |
Chr.6:36594313-36604467 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF3 | NM_003017.4 | NP_003008.1 | ||
| NR_036610.1 | ||||
| AB451229.1 | ||||
| AB451352.1 | ||||
| AF107405.1 | AAD44523.1 | |||
| AK304097.1 | ||||
| AK312553.1 | ||||
| BC000914.1 | AAH00914.1 | |||
| BC069018.1 | AAH69018.1 | |||
| BT007017.1 | AAP35663.1 | |||
| DC398250.1 | ||||
| L10838.1 | AAA36648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map