Assay Details
Target Gene Details
Entrez Gene ID: | 91749 |
Gene Name: | major facilitator superfamily domain containing 4B |
Gene Aliases: |
KIAA1919, NaGLT1 |
Location: |
Chr.6:111259279-111280005 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFSD4B | NM_153369.2 | 4 | 1219 | NP_699200.2 |
| XM_017011483.1 | 3 | 1102 | XP_016866972.1 | |
| AB067506.1 | 1 | 454 | BAB67812.1 | |
| AK093136.1 | 4 | 1178 | ||
| AK292735.1 | 4 | 1148 | ||
| AK315016.1 | 4 | 1141 | ||
| BC036115.1 | 4 | 1128 | AAH36115.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020627 | Chr.6:110982539 - 111302511 on Build GRCh38 | Gain |
|
| nsv1028349 | Chr.6:111184772 - 111371598 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map