Assay Details
Target Gene Details
Entrez Gene ID: | 5934 |
Gene Name: | RB transcriptional corepressor like 2 |
Gene Aliases: |
P130, Rb2 |
Location: |
Chr.16:53434420-53491648 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBL2 | NM_001323608.1 | 12 | 1753 | NP_001310537.1 |
| NM_001323609.1 | 12 | 1753 | NP_001310538.1 | |
| NM_001323610.1 | 12 | 1753 | NP_001310539.1 | |
| NM_001323611.1 | 12 | 1502 | NP_001310540.1 | |
| NM_005611.3 | 12 | 1734 | NP_005602.3 | |
| XM_011523253.2 | 10 | 1127 | XP_011521555.1 | |
| XM_017023513.1 | 9 | 1291 | XP_016879002.1 | |
| AK300047.1 | 12 | 1472 | ||
| AK304283.1 | 9 | 1201 | ||
| AK307881.1 | 12 | 1711 | ||
| BC034490.1 | 12 | 1724 | AAH34490.1 | |
| BX537767.1 | 11 | 1315 | CAD97830.1 | |
| S67171.1 | 12 | 1644 | AAB29227.1 | |
| X74594.1 | 12 | 1445 | CAA52671.1 | |
| X76061.1 | 12 | 1685 | CAA53661.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs140896175] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map