Assay Details
Target Gene Details
Entrez Gene ID: | 6331 |
Gene Name: | sodium voltage-gated channel alpha subunit 5 |
Gene Aliases: |
CDCD2, CMD1E, CMPD2, HB1, HB2, HBBD, HH1, ICCD, IVF, LQT3, Nav1.5, PFHB1, SSS1, VF1 |
Location: |
Chr.3:38548061-38649673 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN5A | NM_000335.4 | 28 | 5757 | NP_000326.2 |
| NM_001099404.1 | 28 | 5760 | NP_001092874.1 | |
| NM_001099405.1 | 27 | 5706 | NP_001092875.1 | |
| NM_001160160.1 | 28 | 5662 | NP_001153632.1 | |
| NM_001160161.1 | 27 | 5599 | NP_001153633.1 | |
| NM_198056.2 | 28 | 5760 | NP_932173.1 | |
| XM_011533991.2 | 28 | 5752 | XP_011532293.1 | |
| XM_017007017.1 | 27 | 5593 | XP_016862506.1 | |
| AB158469.2 | 27 | 5608 | BAD12084.1 | |
| AB158470.2 | 26 | 5446 | BAD12085.1 | |
| AB208866.1 | 18 | 4247 | BAD92103.1 | |
| AF482988.1 | 27 | 5572 | AAO91669.1 | |
| AY038064.1 | 27 | 5621 | AAK74065.1 | |
| AY148488.1 | 27 | 5563 | AAN61120.1 | |
| BC140813.1 | 27 | 5492 | ||
| BC144621.1 | 27 | 5492 | ||
| EF092293.1 | ||||
| EF629346.1 | 27 | 5608 | ||
| EF629347.1 | 26 | 5554 | ||
| KC858891.1 | 26 | 5509 | ||
| M77235.1 | 28 | 5716 | AAA58644.1 |
More Information
Set Membership: |
|

Genomic Map