Assay Details
Target Gene Details
Entrez Gene ID: | 29902 |
Gene Name: | family with sequence similarity 216 member A |
Gene Aliases: |
C12orf24, HSU79274 |
Location: |
Chr.12:110468424-110490387 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM216A | NM_013300.2 | 1 | 402 | NP_037432.2 |
| XM_005253875.4 | XP_005253932.1 | |||
| AK225731.1 | 1 | 393 | ||
| AK296280.1 | 1 | 414 | ||
| AK297684.1 | ||||
| AK308802.1 | ||||
| AK309603.1 | ||||
| AU129789.1 | 1 | 402 | ||
| BC020967.1 | 1 | 218 | AAH20967.1 | |
| U79274.1 | 1 | 369 | AAB50215.1 |
Target Gene Details
Entrez Gene ID: | 51184 |
Gene Name: | GPN-loop GTPase 3 |
Gene Aliases: |
ATPBD1C |
Location: |
Chr.12:110452486-110468835 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPN3 | XM_005253896.4 | XP_005253953.3 |
More Information
Set Membership: |
|

Genomic Map