Assay Details
Target Gene Details
Entrez Gene ID: | 388815 |
Gene Name: | mir-99a-let-7c cluster host gene |
Gene Aliases: |
C21orf34, C21orf35, LINC00478, MONC |
Location: |
Chr.21:16070488-16631727 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR99AHG | NR_027790.3 | 2 | 83 | |
| NR_111004.2 | 1 | 56 | ||
| NR_111005.2 | 1 | 56 | ||
| NR_136541.1 | 2 | 136 | ||
| NR_136542.1 | 2 | 136 | ||
| AF486622.1 | ||||
| AK095614.1 | 2 | 83 | ||
| CN313455.1 | 1 | 56 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527854 | Chr.21:16028596 - 16087187 on Build GRCh38 | Loss |
|

Genomic Map