Assay Details
Target Gene Details
Entrez Gene ID: | 94234 |
Gene Name: | forkhead box Q1 |
Gene Aliases: |
HFH1 |
Location: |
Chr.6:1312440-1314758 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXQ1 | NM_033260.3 | NP_150285.3 | ||
| BC053850.1 | AAH53850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823393 | Chr.6:1307835 - 1321749 on Build GRCh38 | Gain |
|
| nsv1017614 | Chr.6:1248415 - 1423267 on Build GRCh38 | Loss |
|
| esv3576090 | Chr.6:1192187 - 1390975 on Build GRCh38 | Gain |
|
| dgv5910n100 | Chr.6:1268949 - 1328788 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map