Assay Details
Target Gene Details
Entrez Gene ID: | 6047 |
Gene Name: | ring finger protein 4 |
Gene Aliases: |
RES4-26, SLX5, SNURF |
Location: |
Chr.4:2469068-2515859 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF4 | NM_001185010.2 | NP_001171939.1 | ||
| NM_002938.4 | NP_002929.1 | |||
| AB000468.1 | BAA19122.1 | |||
| AK307372.1 | ||||
| AK309509.1 | ||||
| AK312534.1 | ||||
| DA342512.1 | ||||
| U95140.1 | AAC52022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain |
|
| nsv1010352 | Chr.4:2431472 - 2480236 on Build GRCh38 | Gain |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv518472 | Chr.4:2346678 - 2515720 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map