Assay Details
Target Gene Details
Entrez Gene ID: | 10713 |
Gene Name: | ubiquitin specific peptidase 39 |
Gene Aliases: |
65K, CGI-21, HSPC332, SAD1, SNRNP65 |
Location: |
Chr.2:85602842-85649284 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP39 | NM_001256725.1 | 3 | 489 | NP_001243654.1 |
| NM_001256726.1 | 3 | 489 | NP_001243655.1 | |
| NM_001256727.1 | 3 | 374 | NP_001243656.1 | |
| NM_001256728.1 | NP_001243657.1 | |||
| NM_006590.3 | 3 | 489 | NP_006581.2 | |
| NR_046347.1 | 3 | 357 | ||
| XM_006711922.1 | 3 | 357 | XP_006711985.1 | |
| XM_006711923.2 | 3 | 436 | XP_006711986.1 | |
| XM_011532487.1 | 3 | 422 | XP_011530789.1 | |
| XM_011532488.1 | 3 | 355 | XP_011530790.1 | |
| XM_017003182.1 | 3 | 426 | XP_016858671.1 | |
| AF130096.1 | 2 | 78 | AAG35521.1 | |
| AF132955.1 | 3 | 393 | AAD27730.1 | |
| AF353989.1 | 3 | 396 | AAK49524.1 | |
| AK001047.1 | 3 | 387 | ||
| AK056334.1 | 3 | 338 | ||
| AK129872.1 | 4 | 370 | ||
| AK222852.1 | 3 | 392 | BAD96572.1 | |
| AK289451.1 | 3 | 443 | ||
| AK295257.1 | ||||
| AK302236.1 | 3 | 351 | ||
| AK309485.1 | 4 | 292 | ||
| AL117416.1 | ||||
| AL519827.3 | 3 | 439 | ||
| BC001384.2 | 3 | 378 | AAH01384.2 | |
| BC067273.1 | 3 | 417 | AAH67273.1 | |
| BX462623.2 | 3 | 422 | ||
| DA548921.1 | 3 | 358 | ||
| DB124294.1 | 3 | 489 | ||
| DB444135.1 | 3 | 376 |
More Information
Set Membership: |
|

Genomic Map