Assay Details
Target Gene Details
Entrez Gene ID: | 10089 |
Gene Name: | potassium two pore domain channel subfamily K member 7 |
Gene Aliases: |
K2p7.1, TWIK3 |
Location: |
Chr.11:65592855-65595996 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK7 | NM_005714.1 | 2 | 956 | NP_005705.1 |
| NM_033347.1 | NP_203133.1 | |||
| NM_033348.1 | NP_203134.1 | |||
| NM_033455.1 | NP_258416.1 | |||
| AF110522.1 | AAD29578.1 | |||
| AF110523.1 | AAD29579.1 | |||
| AF110524.1 | 2 | 956 | AAD29580.1 | |
| BC103809.1 | AAI03810.1 | |||
| BC103810.2 | AAI03811.1 | |||
| BC103811.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761680 | Chr.11:65576857 - 65623846 on Build GRCh38 | Gain |
|
| dgv1969n54 | Chr.11:65568234 - 65663889 on Build GRCh38 | Loss |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map