Assay Details
Target Gene Details
Entrez Gene ID: | 100507291 |
Gene Name: | uncharacterized LOC100507291 |
Gene Aliases: |
- |
Location: |
Chr.3:139389803-139583319 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507291 | NR_121609.1 | |||
| DA364121.1 | ||||
| DB034152.1 |
Target Gene Details
Entrez Gene ID: | 5948 |
Gene Name: | retinol binding protein 2 |
Gene Aliases: |
CRABP-II, CRBP2, CRBPII, RBPC2 |
Location: |
Chr.3:139452884-139476510 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP2 | NM_004164.2 | 4 | 459 | NP_004155.2 |
| AI076320.1 | 1 | 242 | ||
| AK291978.1 | ||||
| AW772327.1 | 1 | 230 | ||
| BC069296.1 | AAH69296.1 | |||
| BC069361.1 | AAH69361.1 | |||
| BC069396.1 | AAH69396.1 | |||
| BC069424.1 | AAH69424.1 | |||
| BC069447.1 | AAH69447.1 | |||
| BC069469.1 | AAH69469.1 | |||
| BC069513.1 | AAH69513.1 | |||
| BC069522.1 | AAH69522.1 | |||
| U13831.1 | AAC50162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1000519 | Chr.3:139440458 - 139487403 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map