Assay Details
Target Gene Details
Entrez Gene ID: | 51314 |
Gene Name: | NME/NM23 family member 8 |
Gene Aliases: |
CILD6, HEL-S-99, NM23-H8, SPTRX2, TXNDC3, sptrx-2 |
Location: |
Chr.7:37848597-37900401 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME8 | NM_016616.4 | 11 | 1111 | NP_057700.3 |
| AF202051.2 | 10 | 840 | AAF20909.2 | |
| AF305596.1 | 10 | 809 | AAN04258.1 | |
| AK313599.1 | 10 | 851 | ||
| BC036816.1 | 11 | 1111 | AAH36816.1 | |
| GQ472221.1 | 11 | 1111 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8074 | Chr.7:37825738 - 38170485 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map