Assay Details
Target Gene Details
Entrez Gene ID: | 284111 |
Gene Name: | solute carrier family 13 member 5 |
Gene Aliases: |
EIEE25, NACT, mIndy |
Location: |
Chr.17:6684713-6713421 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC13A5 | NM_001143838.2 | 11 | 2938 | NP_001137310.1 |
| NM_001284509.1 | 12 | 3025 | NP_001271438.1 | |
| NM_001284510.1 | 11 | 2947 | NP_001271439.1 | |
| NM_177550.4 | 12 | 3076 | NP_808218.1 | |
| AK124317.1 | 1 | 4432 | ||
| AK127797.1 | 12 | 2973 | ||
| AK172779.1 | 5 | 1929 | BAD18760.1 | |
| AK172785.1 | 12 | 3021 | BAD18766.1 | |
| AY151833.1 | 12 | 3000 | AAN86530.1 | |
| BX647354.1 | 1 | 5195 | ||
| CR749584.1 | 7 | 2190 | CAH18382.1 | |
| H29568.1 | 1 | 241 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574278 | Chr.17:5546345 - 6722066 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map