Assay Details
Target Gene Details
Entrez Gene ID: | 25930 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 23 |
Gene Aliases: |
HD-PTP, HDPTP, PTP-TD14 |
Location: |
Chr.3:47380982-47413441 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPN23 | NM_001304482.1 | 1 | 167 | NP_001291411.1 |
| NM_015466.3 | 1 | 167 | NP_056281.1 | |
| XM_005265031.2 | 1 | 148 | XP_005265088.2 | |
| AB025194.1 | 1 | 115 | BAB19280.1 | |
| AB040904.2 | 1 | 167 | ||
| AF290614.1 | 1 | 113 | AAK28025.1 | |
| AK055157.1 | 1 | 138 | ||
| AK289502.1 | 1 | 116 | ||
| AK295877.1 | 1 | 87 | ||
| AK295888.1 | 1 | 135 | ||
| AK299906.1 | 1 | 138 | ||
| BC089042.1 | 1 | 85 | AAH89042.1 | |
| DA093189.1 | 1 | 148 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3802 | Chr.3:47355184 - 47400461 on Build GRCh38 | Deletion |
|
| nsv3803 | Chr.3:47372642 - 47404507 on Build GRCh38 | Insertion |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map