Assay Details
Target Gene Details
Entrez Gene ID: | 222662 |
Gene Name: | lipoma HMGIC fusion partner-like 5 |
Gene Aliases: |
DFNB67, TMHS, dJ510O8.8 |
Location: |
Chr.6:35805294-35824075 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHFPL5 | NM_182548.3 | 4 | 1619 | NP_872354.1 |
| AK126841.1 | 4 | 1534 | ||
| BC028630.2 | 4 | 1619 | AAH28630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821646 | Chr.6:35636651 - 35834817 on Build GRCh38 | Loss |
|
| esv2759420 | Chr.6:35495964 - 35838727 on Build GRCh38 | Gain |
|
| nsv428143 | Chr.6:35669306 - 35838727 on Build GRCh38 | Gain |
|
| esv3890842 | Chr.6:35678658 - 35837356 on Build GRCh38 | Gain |
|
| esv3608689 | Chr.6:35664221 - 35838480 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map