Assay Details
Target Gene Details
Entrez Gene ID: | 64343 |
Gene Name: | 5-azacytidine induced 2 |
Gene Aliases: |
AZ2, NAP1, TILP |
Location: |
Chr.3:28322353-28349129 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AZI2 | NM_001271650.1 | 9 | 3107 | NP_001258579.1 |
| NM_022461.4 | 8 | 3043 | NP_071906.1 | |
| XM_005265392.2 | 9 | 3130 | XP_005265449.1 | |
| XM_011534009.2 | 8 | 2583 | XP_011532311.1 | |
| BM982133.1 | 1 | 373 | ||
| BX648471.1 | 9 | 2771 |
Target Gene Details
Entrez Gene ID: | 152100 |
Gene Name: | C-X9-C motif containing 1 |
Gene Aliases: |
C3orf68 |
Location: |
Chr.3:28241588-28322874 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMC1 | XM_011533402.2 | 6 | 3740 | XP_011531704.1 |
| BX648671.1 | 4 | 3590 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3568668 | Chr.3:28319282 - 28351354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map