Assay Details
Target Gene Details
Entrez Gene ID: | 140831 |
Gene Name: | zinc finger SWIM-type containing 3 |
Gene Aliases: |
C20orf164, PPP1R174 |
Location: |
Chr.20:45857581-45879131 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSWIM3 | NM_080752.3 | 1 | 339 | NP_542790.2 |
| NR_037628.1 | 1 | 339 | ||
| AK056641.1 | 1 | 304 | BAB71239.1 | |
| AK302767.1 | 1 | 303 | ||
| AK316214.1 | 1 | 302 | ||
| BC027611.2 | 1 | 296 | AAH27611.1 | |
| BX385024.2 | 1 | 297 | ||
| DA520427.1 | 1 | 339 | ||
| DC363945.1 | 1 | 302 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517296 | Chr.20:45854586 - 45965936 on Build GRCh38 | Loss |
|
| nsv458993 | Chr.20:45842587 - 45880235 on Build GRCh38 | Loss |
|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map