Assay Details
Target Gene Details
Entrez Gene ID: | 55323 |
Gene Name: | La ribonucleoprotein domain family member 6 |
Gene Aliases: |
ACHN |
Location: |
Chr.15:70831524-70854265 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP6 | NM_001286679.1 | 3 | 1414 | NP_001273608.1 |
| NM_018357.3 | 3 | 1679 | NP_060827.2 | |
| AF443828.1 | 3 | 1572 | AAN76710.1 | |
| AF443829.1 | 3 | 1572 | AAN76711.1 | |
| AI376944.1 | ||||
| AK002058.1 | 3 | 1573 | BAA92061.1 | |
| AK130092.1 | 5 | 1065 | ||
| AL833876.1 | 3 | 1533 | CAD38733.2 | |
| BC006082.1 | 3 | 1526 | AAH06082.1 | |
| BC009446.1 | 3 | 1526 | AAH09446.1 | |
| BC014018.2 | 3 | 1530 | AAH14018.1 |
More Information
Set Membership: |
|

Genomic Map