Assay Details
Target Gene Details
Entrez Gene ID: | 51151 |
Gene Name: | solute carrier family 45 member 2 |
Gene Aliases: |
1A1, AIM1, MATP, OCA4, SHEP5 |
Location: |
Chr.5:33944616-33984675 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC45A2 | NM_001012509.3 | NP_001012527.1 | ||
| NM_016180.4 | NP_057264.3 | |||
| AF172849.1 | AAD51812.1 | |||
| BC064405.1 | AAH64405.1 | |||
| BU166522.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv597792 | Chr.5:33922453 - 33977730 on Build GRCh38 | Loss |
|
| nsv830255 | Chr.5:33929507 - 34109160 on Build GRCh38 | Loss |
|
| esv3894173 | Chr.5:33945653 - 33958854 on Build GRCh38 | Loss |
|
| nsv462115 | Chr.5:33935838 - 33968785 on Build GRCh38 | Loss |
|
| nsv1019937 | Chr.5:33921280 - 33990689 on Build GRCh38 | Loss |
|
| nsv522715 | Chr.5:33319157 - 34037875 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map