Assay Details
Target Gene Details
Entrez Gene ID: | 2635 |
Gene Name: | guanylate binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.1:89006677-89022880 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP3 | NM_001319179.1 | NP_001306108.1 | ||
| NM_001319180.1 | NP_001306109.1 | |||
| NM_001319181.1 | NP_001306110.1 | |||
| NM_018284.2 | NP_060754.2 | |||
| NR_135007.1 | ||||
| XM_005270743.2 | XP_005270800.1 | |||
| XM_006710574.2 | XP_006710637.1 | |||
| XM_011541233.1 | XP_011539535.1 | |||
| XM_011541234.1 | XP_011539536.1 | |||
| XM_017001013.1 | XP_016856502.1 | |||
| XM_017001014.1 | XP_016856503.1 | |||
| AK001823.1 | ||||
| AK309096.1 | ||||
| AL136680.1 | CAB66615.1 | |||
| BC017992.1 | ||||
| BC140837.1 | ||||
| CR936755.1 | ||||
| CR936758.1 | ||||
| JF927951.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546793 | Chr.1:89011781 - 89013290 on Build GRCh38 | Loss |
|
| esv3565560 | Chr.1:89009936 - 89013002 on Build GRCh38 | Deletion |
|
| nsv946055 | Chr.1:89010812 - 89014409 on Build GRCh38 | Duplication |
|
| esv22437 | Chr.1:89010736 - 89012843 on Build GRCh38 | Gain |
|
| dgv361n54 | Chr.1:89011229 - 89012655 on Build GRCh38 | Gain |
|
| nsv546786 | Chr.1:89010744 - 89012749 on Build GRCh38 | Loss |
|
| dgv55n67 | Chr.1:89010510 - 89012942 on Build GRCh38 | Loss |
|
| nsv819062 | Chr.1:89010401 - 89013391 on Build GRCh38 | Gain |
|
| nsv10361 | Chr.1:89010060 - 89013363 on Build GRCh38 | Gain |
|
| esv3586751 | Chr.1:89010247 - 89012930 on Build GRCh38 | Loss |
|
| esv2667765 | Chr.1:89010225 - 89012941 on Build GRCh38 | Deletion |
|
| nsv1071104 | Chr.1:89010116 - 89013117 on Build GRCh38 | Deletion |
|
| esv2758948 | Chr.1:88946385 - 89095040 on Build GRCh38 | Gain |
|
| esv2714551 | Chr.1:89010106 - 89012914 on Build GRCh38 | Deletion |
|
| dgv62e212 | Chr.1:89009426 - 89012283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map