Assay Details
Target Gene Details
Entrez Gene ID: | 84720 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class O |
Gene Aliases: |
HPMRS2 |
Location: |
Chr.9:35085493-35096601 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGO | NM_001201484.1 | 3 | 915 | NP_001188413.1 |
| NM_032634.3 | 2 | 817 | NP_116023.2 | |
| NM_152850.3 | 2 | 463 | NP_690577.2 | |
| XM_005251619.3 | 2 | 442 | XP_005251676.1 | |
| XM_017015222.1 | 3 | 934 | XP_016870711.1 | |
| XM_017015223.1 | 2 | 844 | XP_016870712.1 | |
| XM_017015224.1 | 2 | 1108 | XP_016870713.1 | |
| AK074064.1 | 2 | 438 | BAB84890.1 | |
| AK090433.1 | 1 | 1406 | BAC03414.1 | |
| AL833956.1 | 2 | 854 | CAD38806.1 | |
| AY358472.1 | 1 | 423 | AAQ88836.1 | |
| BC029271.1 | 1 | 88 | AAH29271.1 | |
| BC036916.1 | 1 | 428 | AAH36916.1 | |
| BC065282.1 | 3 | 586 | AAH65282.1 | |
| BI818120.1 | 2 | 457 | ||
| CD387014.1 | 2 | 454 | ||
| CR991745.1 | 2 | 463 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030303 | Chr.9:35084357 - 35506646 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116248521] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map