Assay Details
Target Gene Details
Entrez Gene ID: | 54460 |
Gene Name: | mitochondrial ribosomal protein S21 |
Gene Aliases: |
MDS016, MRP-S21, RPMS21 |
Location: |
Chr.1:150293842-150308979 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS21 | NM_018997.3 | NP_061870.1 | ||
| NM_031901.5 | NP_114107.1 | |||
| AA977333.1 | ||||
| AB049955.1 | BAB41008.1 | |||
| AF182417.1 | AAG14953.1 | |||
| AK289467.1 | ||||
| BC004566.2 | AAH04566.2 | |||
| BC006554.2 | AAH06554.1 | |||
| BG037004.1 | ||||
| BI755132.1 | ||||
| HY161691.1 | ||||
| U79258.1 | AAB50204.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009508 | Chr.1:150238763 - 150298405 on Build GRCh38 | Loss |
|
| nsv1011223 | Chr.1:150256810 - 150306949 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map