Assay Details
Target Gene Details
Entrez Gene ID: | 151354 |
Gene Name: | family with sequence similarity 84 member A |
Gene Aliases: |
NSE1, PP11517 |
Location: |
Chr.2:14632686-14640046 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM84A | NM_145175.2 | 2 | 1744 | NP_660158.2 |
| AF370404.1 | 2 | 1690 | AAQ15240.1 | |
| AJ417080.1 | 2 | 1698 | CAD10038.1 | |
| AK092654.1 | 3 | 1544 | BAC03938.1 | |
| BC026346.1 | 1 | 1738 | AAH26346.1 | |
| BC052284.1 | 2 | 1693 | AAH52284.1 |
Target Gene Details
Entrez Gene ID: | 653602 |
Gene Name: | uncharacterized LOC653602 |
Gene Aliases: |
- |
Location: |
Chr.2:14635091-14650819 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC653602 | NM_001291410.1 | 1 | 345 | NP_001278339.1 |
| AK092711.1 | 1 | 345 | BAC03955.1 |

Genomic Map