Assay Details
Target Gene Details
Entrez Gene ID: | 84617 |
Gene Name: | tubulin beta 6 class V |
Gene Aliases: |
HsT1601, TUBB-5 |
Location: |
Chr.18:12307669-12329826 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB6 | NM_001303524.1 | 5 | 1829 | NP_001290453.1 |
| NM_001303525.1 | NP_001290454.1 | |||
| NM_001303526.1 | 3 | 1820 | NP_001290455.1 | |
| NM_001303527.1 | 3 | 1822 | NP_001290456.1 | |
| NM_001303528.1 | 5 | 2047 | NP_001290457.1 | |
| NM_001303529.1 | 4 | 2189 | NP_001290458.1 | |
| NM_001303530.1 | 5 | 2398 | NP_001290459.1 | |
| NM_032525.2 | 4 | 1931 | NP_115914.1 | |
| AK001295.1 | 4 | 1749 | ||
| AK022340.1 | 4 | 1749 | BAB14016.1 | |
| AK092677.1 | 3 | 2086 | ||
| AK098440.1 | 4 | 1749 | ||
| BC002654.1 | 4 | 1742 | AAH02654.1 | |
| CD364877.1 | 1 | 148 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060808 | Chr.18:12302793 - 12514418 on Build GRCh38 | Gain |
|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map