Assay Details
Target Gene Details
Entrez Gene ID: | 23411 |
Gene Name: | sirtuin 1 |
Gene Aliases: |
SIR2, SIR2L1, SIR2alpha |
Location: |
Chr.10:67884669-67918390 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRT1 | NM_001142498.1 | NP_001135970.1 | ||
| NM_012238.4 | NP_036370.2 | |||
| AF083106.2 | AAD40849.2 | |||
| AF235040.1 | AAG38486.1 | |||
| AK289743.1 | ||||
| AL136741.1 | ||||
| BC012499.1 | AAH12499.1 | |||
| DB162635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551433 | Chr.10:67805187 - 67937238 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map