Assay Details
Target Gene Details
Entrez Gene ID: | 1509 |
Gene Name: | cathepsin D |
Gene Aliases: |
CLN10, CPSD, HEL-S-130P |
Location: |
Chr.11:1752752-1763992 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSD | NM_001909.4 | 9 | 1848 | NP_001900.1 |
| AK130178.1 | ||||
| BC016320.2 | 9 | 1760 | AAH16320.1 | |
| BM976200.1 | 1 | 294 | ||
| EU794598.1 | 9 | 1760 | ||
| M11233.1 | 9 | 1766 | AAB59529.1 | |
| X05344.1 | 9 | 1717 | CAA28955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832049 | Chr.11:1669923 - 1817378 on Build GRCh38 | Loss |
|
| dgv1564n54 | Chr.11:1752247 - 1785884 on Build GRCh38 | Loss |
|
| nsv7642 | Chr.11:1749402 - 1784428 on Build GRCh38 | Insertion |
|
| nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map