Assay Details
Target Gene Details
Entrez Gene ID: | 2100 |
Gene Name: | estrogen receptor 2 |
Gene Aliases: |
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2 |
Location: |
Chr.14:64226712-64338631 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESR2 | NM_001040275.1 | NP_001035365.1 | ||
| NM_001214902.1 | NP_001201831.1 | |||
| NM_001271876.1 | NP_001258805.1 | |||
| NM_001271877.1 | 6 | 1272 | NP_001258806.1 | |
| NM_001291712.1 | NP_001278641.1 | |||
| NM_001291723.1 | NP_001278652.1 | |||
| NM_001437.2 | 9 | 1923 | NP_001428.1 | |
| NR_073496.1 | ||||
| NR_073497.1 | 7 | 1423 | ||
| XM_017021079.1 | 10 | 2587 | XP_016876568.1 | |
| XM_017021080.1 | 10 | 3396 | XP_016876569.1 | |
| XM_017021081.1 | 10 | 2492 | XP_016876570.1 | |
| XM_017021082.1 | 10 | 2344 | XP_016876571.1 | |
| XM_017021083.1 | 9 | 1761 | XP_016876572.1 | |
| XM_017021084.1 | XP_016876573.1 | |||
| AB006589.1 | BAA31966.1 | |||
| AB006590.1 | 8 | 1553 | BAA24953.1 | |
| AB209620.1 | 7 | 3197 | BAD92857.1 | |
| AF051427.1 | 9 | 1873 | AAC05985.1 | |
| AF051428.1 | AAC05751.1 | |||
| AF060555.1 | AAC15234.1 | |||
| AF061054.1 | AAC39784.1 | |||
| AF061055.1 | AAC39785.1 | |||
| AF074599.1 | 6 | 1010 | AAC25603.1 | |
| AF124790.1 | AAD32580.1 | |||
| AK292370.1 | ||||
| AY438022.1 | 7 | 1192 | ||
| DQ777076.1 | ||||
| DQ838582.1 | ||||
| DQ838583.1 | ||||
| HQ692820.1 | 8 | 1455 | ||
| HQ692821.1 | ||||
| KJ534833.1 | 8 | 1455 | ||
| X99101.1 | 8 | 1314 | CAA67555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050378 | Chr.14:64219825 - 64236294 on Build GRCh38 | Gain |
|
| nsv564905 | Chr.14:63953695 - 64287643 on Build GRCh38 | Gain |
|
| esv3634746 | Chr.14:64169924 - 64294015 on Build GRCh38 | Loss |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map