Assay Details
Target Gene Details
Entrez Gene ID: | 125228 |
Gene Name: | family with sequence similarity 210 member A |
Gene Aliases: |
C18orf19, HsT2329 |
Location: |
Chr.18:13663347-13726592 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM210A | NM_001098801.1 | 5 | 1289 | NP_001092271.1 |
| NM_152352.3 | 4 | 1149 | NP_689565.2 | |
| AK055618.1 | 5 | 1245 | BAB70973.1 | |
| AK309506.1 | 2 | 536 | ||
| AK309819.1 | 2 | 423 | ||
| BU189118.1 | ||||
| CB987819.1 | ||||
| DA573992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576529 | Chr.18:13606185 - 14150725 on Build GRCh38 | Gain |
|

Genomic Map