Assay Details
Target Gene Details
Entrez Gene ID: | 6821 |
Gene Name: | sulfite oxidase |
Gene Aliases: |
- |
Location: |
Chr.12:55996776-56005525 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUOX | NM_000456.2 | NP_000447.2 | ||
| NM_001032386.1 | NP_001027558.1 | |||
| NM_001032387.1 | NP_001027559.1 | |||
| XM_005269112.1 | XP_005269169.1 | |||
| XM_017019905.1 | XP_016875394.1 | |||
| XM_017019907.1 | XP_016875396.1 | |||
| AK309669.1 | ||||
| AK312763.1 | ||||
| AU121557.1 | ||||
| AU126450.1 | ||||
| BC065193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832424 | Chr.12:55881442 - 56073133 on Build GRCh38 | Loss |
|
| nsv1051961 | Chr.12:55868436 - 56235643 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map